Tisk…

4-(2-Bromopyridin-4-yl)piperazine, N1-BOC protected

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Bromopyridin-4-yl)-4-(tert-butoxycarbonyl)piperazine, tert-Butyl 4-(2-bromopyridin-4-yl)piperazine-1-carboxylate

OR302092-1G OR302092-250MG OR302092-5G
APOSOR302092-1GEA 13030 CZK
APOSOR302092-1G APOSOR302092-250MG APOSOR302092-5G
4-(2-Bromopyridin-4-yl)piperazine, N1-BOC protected
4-(2-Bromopyridin-4-yl)piperazine, N1-BOC protected
Vzorec: C₁₄H₂₀BrN₃O₂
MW: 342,231 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447575
CAS číslo: 1266118-96-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR