Tisk…

Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate

Dodavatel: Apollo Scientific

OR302059-1G OR302059-5G OR302059-250MG
APOSOR302059-1GEA 8040 CZK
APOSOR302059-1G APOSOR302059-5G APOSOR302059-250MG
Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate
Ethyl 5-amino-1-(pyrimidin-2-yl)-1H-pyrazole-4-carboxylate
Vzorec: C₁₀H₁₁N₅O₂
Storage Temperature: Ambient
MDL Number: MFCD02691657
CAS číslo: 91129-95-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR