Tisk…

5-Bromo-4-chloro-2-methoxypyridine 98%

Dodavatel: Apollo Scientific
Varování

OR301297-1G
APOSOR301297-1GEA 1700 CZK
APOSOR301297-1G
5-Bromo-4-chloro-2-methoxypyridine 98%
5-Bromo-4-chloro-2-methoxypyridine
Vzorec: C₆H₅BrClNO
MW: 222,467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17014994
CAS číslo: 851607-27-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR