Tisk…

4-Bromo-1,3-thiazole-2-carboxylic acid 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromothiazole-2-carboxylic acid

OR301250-1G OR301250-5G
APOSOR301250-1GEA 2030 CZK
APOSOR301250-1G APOSOR301250-5G
4-Bromo-1,3-thiazole-2-carboxylic acid 96%
4-Bromo-1,3-thiazole-2-carboxylic acid
Vzorec: C₄H₂BrNO₂S
MW: 208,033 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09701413
CAS číslo: 88982-82-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR