Tisk…

2,4-Dichloropyrido[3,4-d]pyrimidine 96%

Dodavatel: Apollo Scientific
Varování

OR301218-100MG OR301218-1G OR301218-250MG
APOSOR301218-100MGEA 3480 CZK
APOSOR301218-100MG APOSOR301218-1G APOSOR301218-250MG
2,4-Dichloropyrido[3,4-d]pyrimidine 96%
2,4-Dichloropyrido[3,4-d]pyrimidine
Vzorec: C₇H₃Cl₂N₃
MW: 200,025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699461
CAS číslo: 908240-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR