Tisk…

4-Chlorotetrazolo[1,5-a]quinoxaline 95%

Dodavatel: Apollo Scientific
Varování

OR301213-1G OR301213-250MG
APOSOR301213-1GEA 20470 CZK
APOSOR301213-1G APOSOR301213-250MG
4-Chlorotetrazolo[1,5-a]quinoxaline 95%
4-Chlorotetrazolo[1,5-a]quinoxaline
Vzorec: C₈H₄ClN₅
MW: 205.604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05717322
CAS číslo: 59866-06-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR