Methyl 2-oxopiperidine-4-carboxylate 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR301040-1G
OR301040-5G
APOSOR301040-1GEA
3060
CZK
APOSOR301040-1G
APOSOR301040-5G
Methyl 2-oxopiperidine-4-carboxylate 96%
Methyl 2-oxopiperidine-4-carboxylate
Vzorec:
C₇H₁₁NO₃ Bod tání: 115…127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD17012745 CAS číslo: 25504-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...