Tisk…

1-(N-Boc-amino)-3-cyclopentene

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl cyclopent-3-en-1-ylcarbamate

OR301010-5G OR301010-250MG OR301010-1G
APOSOR301010-5GEA 8830 CZK
APOSOR301010-5G APOSOR301010-250MG APOSOR301010-1G
1-(N-Boc-amino)-3-cyclopentene
1-(N-Boc-amino)-3-cyclopentene
Vzorec: C₁₀H₁₇NO₂
MW: 183,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179403
CAS číslo: 193751-54-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR