Tisk…

3-Methyl-2-(1-piperidin-1-yl)butyronitrile hydrochloride

Dodavatel: Apollo Scientific

OR300895-1G OR300895-5G
APOSOR300895-1GEA 0 CZK
APOSOR300895-1G APOSOR300895-5G
3-Methyl-2-(1-piperidin-1-yl)butyronitrile hydrochloride
3-Methyl-2-(1-piperidin-1-yl)butyronitrile hydrochloride
Vzorec: C₁₀H₁₈N₂·ClH
MW: 202,724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28122844
CAS číslo: 1672675-20-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR