Methyl 2-cyano-3-methylbut-2-enoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR300789-10G
OR300789-1G
OR300789-5G
APOSOR300789-10GEA
16770
CZK
APOSOR300789-10G
APOSOR300789-1G
APOSOR300789-5G
Methyl 2-cyano-3-methylbut-2-enoate
Methyl 2-cyano-3-methylbut-2-enoate
Vzorec:
C₇H₉NO₂ Bod tání: 20…21 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085476 CAS číslo: 6666-75-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...