Tisk…
2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride
Dodavatel: Apollo Scientific
Synonyms: α-(4-Morpholino)-4-chlorophenylacetonitrile hydrochloride
OR300509-10G OR300509-1G OR300509-5G
APOSOR300509-10GEA 0 CZK
APOSOR300509-10G APOSOR300509-1G APOSOR300509-5G
2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride
2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride
Vzorec: C₁₂H₁₃ClN₂O·ClH
MW: 273.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052434
CAS číslo: 1330171-75-9