7-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR300408-1G
OR300408-250MG
APOSOR300408-1GEA
9720
CZK
APOSOR300408-1G
APOSOR300408-250MG
7-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
7-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
Vzorec:
C₆H₃ClIN₃ MW: 279,466 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18633053 CAS číslo: 1268521-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...