1-(5-Bromo-3-methylpyridin-2-yl)ethanone
Dodavatel: Apollo Scientific
OR300405-1G
OR300405-250MG
APOSOR300405-1GEA
20500
CZK
APOSOR300405-1G
APOSOR300405-250MG
1-(5-Bromo-3-methylpyridin-2-yl)ethanone
1-(5-Bromo-3-methylpyridin-2-yl)ethanone
Vzorec:
C₈H₈BrNO MW: 214.059 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18261721 CAS číslo: 1211533-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...