Tisk…

5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine

Dodavatel: Apollo Scientific
Varování

OR300392-1G OR300392-250MG
APOSOR300392-1GEA 7910 CZK
APOSOR300392-1G APOSOR300392-250MG
5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
Vzorec: C₆H₃ClIN₃
MW: 279,466 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18250844
CAS číslo: 1260666-29-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR