Tisk…

7-Bromopyrido[3,4-b]pyrazine

Dodavatel: Apollo Scientific
Varování

OR300382-1G OR300382-250MG
APOSOR300382-1GEA 10250 CZK
APOSOR300382-1G APOSOR300382-250MG
7-Bromopyrido[3,4-b]pyrazine
7-Bromopyrido[3,4-b]pyrazine
Vzorec: C₇H₄BrN₃
MW: 210,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380148
CAS číslo: 1337880-74-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR