3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR300376-1G
OR300376-250MG
APOSOR300376-1GEA
18490
CZK
APOSOR300376-1G
APOSOR300376-250MG
3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine
3,5-Dibromo-1H-pyrazolo[3,4-b]pyridine
Vzorec:
C₆H₃Br₂N₃ Storage Temperature: Ambient |
MDL Number:
MFCD20923265 CAS číslo: 1352395-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...