Tisk…

3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine

Dodavatel: Apollo Scientific
Varování

OR300374-1G OR300374-250MG
APOSOR300374-1GEA 9360 CZK
APOSOR300374-1G APOSOR300374-250MG
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine
Vzorec: C₇H₄BrClN₂
MW: 231,477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007847
CAS číslo: 947238-42-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR