2-Methoxypyridine-3,4-diamine
Dodavatel: Apollo Scientific
|
Varování
|
OR300373-1G
OR300373-500MG
APOSOR300373-1GEA
14490
CZK
APOSOR300373-1G
APOSOR300373-500MG
2-Methoxypyridine-3,4-diamine
2-Methoxypyridine-3,4-diamine
Vzorec:
C₆H₉N₃O MW: 139,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10574697 CAS číslo: 33631-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...