Tisk…

3-Bromoquinolin-5-amine

Dodavatel: Apollo Scientific
Varování

OR300363-1G OR300363-250MG
APOSOR300363-1GEA 4040 CZK
APOSOR300363-1G APOSOR300363-250MG
3-Bromoquinolin-5-amine
3-Bromoquinolin-5-amine
Vzorec: C₉H₇BrN₂
MW: 223,069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20327979
CAS číslo: 116632-57-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR