tert-Butyl (5-bromo-6-chloropyridin-3-yl)methylcarbamate
Dodavatel: Apollo Scientific
|
Varování
|
OR300320-100MG
OR300320-250MG
APOSOR300320-100MGEA
5200
CZK
APOSOR300320-100MG
APOSOR300320-250MG
tert-Butyl (5-bromo-6-chloropyridin-3-yl)methylcarbamate
tert-Butyl (5-bromo-6-chloropyridin-3-yl)methylcarbamate
Vzorec:
C₁₁H₁₄BrClN₂O₂ MW: 321,598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31621104 CAS číslo: 2231673-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...