Tisk…

7-Bromo-2,3-dihydro-1H-carbazol-4(9H)-one

Dodavatel: Apollo Scientific
Varování

OR300315-100MG OR300315-250MG
APOSOR300315-100MGEA 5100 CZK
APOSOR300315-100MG APOSOR300315-250MG
7-Bromo-2,3-dihydro-1H-carbazol-4(9H)-one
7-Bromo-2,3-dihydro-1H-carbazol-4(9H)-one
Vzorec: C₁₂H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD27937964
CAS číslo: 1260493-98-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR