7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one
Dodavatel: Apollo Scientific
|
Varování
|
OR300314-1G
OR300314-250MG
APOSOR300314-1GEA
41330
CZK
APOSOR300314-1G
APOSOR300314-250MG
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one
7-Bromo-2,3-dihydro-2,2-dimethyl-1H-carbazol-4(9H)-one
Vzorec:
C₁₄H₁₄BrNO Storage Temperature: Ambient |
MDL Number:
MFCD29765459 CAS číslo: 1301219-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...