Tisk…

2-Bromo-1H-indole-6-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR300308-1G OR300308-250MG
APOSOR300308-1GEA 49260 CZK
APOSOR300308-1G APOSOR300308-250MG
2-Bromo-1H-indole-6-carboxylic acid
2-Bromo-1H-indole-6-carboxylic acid
Vzorec: C₉H₆BrNO₂
MW: 240,053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31621098
CAS číslo: 1782429-68-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR