Tisk…

2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine

Dodavatel: Apollo Scientific
Varování

OR300260-100MG OR300260-250MG
APOSOR300260-100MGEA 16960 CZK
APOSOR300260-100MG APOSOR300260-250MG
2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine
2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine
Vzorec: C₇H₉BrN₂S
MW: 233,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22571362
CAS číslo: 1280704-26-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR