2-Bromo-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR300257-100MG
OR300257-250MG
APOSOR300257-100MGEA
12300
CZK
APOSOR300257-100MG
APOSOR300257-250MG
2-Bromo-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine
2-Bromo-4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine
Vzorec:
C₆H₇BrN₂S MW: 219,102 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22036527 CAS číslo: 1368338-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...