Tisk…

tert-Butyl 3-bromo-5-methylbenzylcarbamate

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (3-bromo-5-methylbenzyl)carbamate

OR300247-1G OR300247-250MG
APOSOR300247-1GEA 17590 CZK
APOSOR300247-1G APOSOR300247-250MG
tert-Butyl 3-bromo-5-methylbenzylcarbamate
tert-Butyl 3-bromo-5-methylbenzylcarbamate
Vzorec: C₁₃H₁₈BrNO₂
MW: 300,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30531038
CAS číslo: 1998726-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR