(3-Bromo-5-methylphenyl)methanamine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR300246-1G
OR300246-250MG
APOSOR300246-1GEA
10910
CZK
APOSOR300246-1G
APOSOR300246-250MG
(3-Bromo-5-methylphenyl)methanamine hydrochloride
(3-Bromo-5-methylphenyl)methanamine hydrochloride
Vzorec:
C₈H₁₀BrN·ClH MW: 236,537 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28893555 CAS číslo: 1646570-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...