5-Iodo-3-phenylpyrazin-2-amine
Dodavatel: Apollo Scientific
|
Varování
|
OR300243-1G
OR300243-250MG
APOSOR300243-1GEA
28440
CZK
APOSOR300243-1G
APOSOR300243-250MG
5-Iodo-3-phenylpyrazin-2-amine
5-Iodo-3-phenylpyrazin-2-amine
Vzorec:
C₁₀H₈IN₃ MW: 297,095 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30528043 CAS číslo: 2090128-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...