3-Iodo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR300188-1G
OR300188-250MG
APOSOR300188-1GEA
11380
CZK
APOSOR300188-1G
APOSOR300188-250MG
3-Iodo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine
3-Iodo-7-methoxy-1-methyl-1H-pyrrolo[2,3-c]pyridine
Vzorec:
C₉H₉IN₂O MW: 288,085 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31536719 CAS číslo: 1627713-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...