Tisk…

2-Aminoquinazolin-4-ol

Dodavatel: Apollo Scientific
Varování

OR300186-1G OR300186-250MG
APOSOR300186-1GEA 5390 CZK
APOSOR300186-1G APOSOR300186-250MG
2-Aminoquinazolin-4-ol
2-Aminoquinazolin-4-ol
Vzorec: C₈H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD18378813
CAS číslo: 20198-19-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR