1-(3-Bromo-5-chloropyridin-2-yl)ethanone
Dodavatel: Apollo Scientific
|
Varování
|
OR300156-1G
OR300156-250MG
APOSOR300156-1GEA
7180
CZK
APOSOR300156-1G
APOSOR300156-250MG
1-(3-Bromo-5-chloropyridin-2-yl)ethanone
1-(3-Bromo-5-chloropyridin-2-yl)ethanone
Vzorec:
C₇H₅BrClNO MW: 234.478 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18255548 CAS číslo: 1256823-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...