Tisk…

3-Chloroisoquinoline-5-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Borono-3-chloro-2-azanaphthalene, 5-Borono-3-chloroisoquinoline, (3-Chloroisoquinolin-5-yl)boronic acid

OR300151-500MG OR300151-100MG
APOSOR300151-500MGEA 12400 CZK
APOSOR300151-500MG APOSOR300151-100MG
3-Chloroisoquinoline-5-boronic acid
3-Chloroisoquinoline-5-boronic acid
Vzorec: C₉H₇BClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22199341
CAS číslo: 1429665-44-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR