Tisk…

Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR300148-1G OR300148-250MG
APOSOR300148-1GEA 15480 CZK
APOSOR300148-1G APOSOR300148-250MG
Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate
Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate
Vzorec: C₁₄H₁₅NO₂S
MW: 261,339 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738347
CAS číslo: 885278-69-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR