Tisk…

5-Bromoisoquinolin-1(2H)-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-1,2-dihydro-1-oxoisoquinoline

OR300119-1G OR300119-250MG OR300119-5G
APOSOR300119-1GEA 3740 CZK
APOSOR300119-1G APOSOR300119-250MG APOSOR300119-5G
5-Bromoisoquinolin-1(2H)-one
5-Bromoisoquinolin-1(2H)-one
Vzorec: C₉H₆BrNO
MW: 224,054 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226915
CAS číslo: 190777-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR