Tisk…

3-Chloroisoquinoline-6-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Carboxy-3-chloroisoquinoline, 6-Carboxy-3-chloro-2-azanaphthalene

OR300082-1G OR300082-250MG
APOSOR300082-1GEA 14880 CZK
APOSOR300082-1G APOSOR300082-250MG
3-Chloroisoquinoline-6-carboxylic acid
3-Chloroisoquinoline-6-carboxylic acid
Vzorec: C₁₀H₆ClNO₂
MW: 207,613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260383
CAS číslo: 1256794-71-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR