7-Chloroisoquinoline
Dodavatel: Apollo Scientific
|
Varování
|
OR300070-250MG
OR300070-1G
APOSOR300070-250MGEA
1620
CZK
APOSOR300070-250MG
APOSOR300070-1G
7-Chloroisoquinoline
7-Chloroisoquinoline
Vzorec:
C₉H₆ClN Storage Temperature: Ambient |
MDL Number:
MFCD00094995 CAS číslo: 34784-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...