Tisk…

1-Amino-8-bromoisoquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Amino-8-bromo-2-azanaphthalene, 8-Bromoisoquinolin-1-amine

OR300063-500MG OR300063-100MG OR300063-250MG
APOSOR300063-500MGEA 21230 CZK
APOSOR300063-500MG APOSOR300063-100MG APOSOR300063-250MG
1-Amino-8-bromoisoquinoline
1-Amino-8-bromoisoquinoline
Vzorec: C₉H₇BrN₂
MW: 223,069 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21365093
CAS číslo: 1337879-85-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR