Tisk…

1-Chloroisoquinoline-5-carbonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Chloro-5-cyanoisoquinoline, 1-Chloro-5-cyano-2-azanaphthalene

OR300058-100MG OR300058-250MG
APOSOR300058-100MGEA 1910 CZK
APOSOR300058-100MG APOSOR300058-250MG
1-Chloroisoquinoline-5-carbonitrile
1-Chloroisoquinoline-5-carbonitrile
Vzorec: C₁₀H₅ClN₂
MW: 188,613 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18250893
CAS číslo: 1231761-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR