Tisk…

1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride

Dodavatel: Apollo Scientific
Varování

OR300040-1G OR300040-250MG
APOSOR300040-1GEA 992 CZK
APOSOR300040-1G APOSOR300040-250MG
1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride
1-(tert-Butylamino)-3-chloropropan-2-ol hydrochloride
Vzorec: C₇H₁₆ClNO·ClH
MW: 202,122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191401
CAS číslo: 41052-94-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR