Tisk…

1-Oxo-1,2,3,4-tetrahydroisoquinoline-6-boronic acid, pinacol ester 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Oxo-1,2,3,4-tetrahydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline, 3,4-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinolin-1(2H)-one, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroisoquinolin-1(2H)-one

OR300011-1G OR300011-250MG
APOSOR300011-1GEA 13230 CZK
APOSOR300011-1G APOSOR300011-250MG
1-Oxo-1,2,3,4-tetrahydroisoquinoline-6-boronic acid, pinacol ester 95+%
1-Oxo-1,2,3,4-tetrahydroisoquinoline-6-boronic acid, pinacol ester
Vzorec: C₁₅H₂₀BNO₃
MW: 273.135 g/mol
Bod tání: 234…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11044671
CAS číslo: 376584-30-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR