1-Chloroisoquinoline-7-boronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Borono-1-chloroisoquinoline, 7-Borono-1-chloro-2-azanaphthalene, (1-Chloroisoquinolin-7-yl)boronic acid
OR300003-250MG
OR300003-500MG
APOSOR300003-250MGEA
4960
CZK
APOSOR300003-250MG
APOSOR300003-500MG
1-Chloroisoquinoline-7-boronic acid
1-Chloroisoquinoline-7-boronic acid
Vzorec:
C₉H₇BClNO₂ MW: 207.421 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13190241 CAS číslo: 370864-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...