Tisk…

2-Bromo-1-[3-(4-chlorophenyl)isoxazol-5-yl]ethan-1-one

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethan-1-one

OR29778-1G OR29778-2.5G
APOSOR29778-1GEA 2820 CZK
APOSOR29778-1G APOSOR29778-2.5G
2-Bromo-1-[3-(4-chlorophenyl)isoxazol-5-yl]ethan-1-one
2-Bromo-1-[3-(4-chlorophenyl)isoxazol-5-yl]ethan-1-one
Vzorec: C₁₁H₇BrClNO₂
MW: 300.536 g/mol
Bod tání: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00662759
CAS číslo: 258506-49-9
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR