Tisk…

2-(2-Thienylmethylidene)hydrazine-1-carbothioamide

Dodavatel: Apollo Scientific

OR29658-10G OR29658-1G OR29658-5G
APOSOR29658-10GEA 24600 CZK
APOSOR29658-10G APOSOR29658-1G APOSOR29658-5G
2-(2-Thienylmethylidene)hydrazine-1-carbothioamide
2-(2-Thienylmethylidene)hydrazine-1-carbothioamide
Vzorec: C₆H₇N₃S₂
MW: 185,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173810
CAS číslo: 5351-91-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR