Tisk…

Methyl 2-[(2,6-dichloro-4-pyridyl)carbonyl]-3-(methylamino)but-2-enoate

Dodavatel: Apollo Scientific

OR29586-1G OR29586-5G
APOSOR29586-1GEA 2900 CZK
APOSOR29586-1G APOSOR29586-5G
Methyl 2-[(2,6-dichloro-4-pyridyl)carbonyl]-3-(methylamino)but-2-enoate
Methyl 2-[(2,6-dichloro-4-pyridyl)carbonyl]-3-(methylamino)but-2-enoate
Vzorec: C₁₂H₁₂Cl₂N₂O₃
MW: 303.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00177420
CAS číslo: 82140-55-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR