5-{3-[(tert-Butylsulphonyl)methyl]-1,2,4-oxadiazol-5-yl}-2-chloropyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR29514-100MG
OR29514-10MG
APOSOR29514-100MGEA
8070
CZK
APOSOR29514-100MG
APOSOR29514-10MG
5-{3-[(tert-Butylsulphonyl)methyl]-1,2,4-oxadiazol-5-yl}-2-chloropyridine
5-{3-[(tert-Butylsulphonyl)methyl]-1,2,4-oxadiazol-5-yl}-2-chloropyridine
Vzorec:
C₁₂H₁₄ClN₃O₃S Storage Temperature: Ambient |
MDL Number:
MFCD02090296 CAS číslo: 680216-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...