5-(Methylsulphonyl)thiophene-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-(Methylsulphonyl)-2-thenoic acid, 2-Carboxy-5-(methylsulphonyl)thiophene
OR29205-1G
OR29205-250MG
APOSOR29205-1GEA
3190
CZK
APOSOR29205-1G
APOSOR29205-250MG
5-(Methylsulphonyl)thiophene-2-carboxylic acid
5-(Methylsulphonyl)thiophene-2-carboxylic acid
Vzorec:
C₆H₆O₄S₂ MW: 206,239 g/mol Bod tání: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173745 CAS číslo: 60166-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...