2,2-Dimethylpentan-3-one
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,2-Dimethyl-3-oxopentane, Ethyl tert-butyl ketone
OR29023-1G
OR29023-250MG
APOSOR29023-1GEA
4570
CZK
APOSOR29023-1G
APOSOR29023-250MG
2,2-Dimethylpentan-3-one
2,2-Dimethylpentan-3-one
Vzorec:
C₇H₁₄O MW: 114,185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00039918 CAS číslo: 564-04-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...