6-Chloro-2,3-dihydro-4H-thiochromen-4-one 1,1-dioxide
Dodavatel: Apollo Scientific
OR29015-1G
OR29015-5G
OR29015-250MG
APOSOR29015-1GEA
3740
CZK
APOSOR29015-1G
APOSOR29015-5G
APOSOR29015-250MG
6-Chloro-2,3-dihydro-4H-thiochromen-4-one 1,1-dioxide
6-Chloro-2,3-dihydro-4H-thiochromen-4-one 1,1-dioxide
Vzorec:
C₉H₇ClO₃S MW: 230,668 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052999 CAS číslo: 90396-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...