2-methylbut-2-enoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR28647-100MG
OR28647-5G
OR28647-25G
APOSOR28647-100MGEA
622
CZK
APOSOR28647-100MG
APOSOR28647-5G
APOSOR28647-25G
2-methylbut-2-enoic acid
2-methylbut-2-enoic acid
Vzorec:
C₅H₈O₂ MW: 100,116 g/mol Bod tání: 44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066864 CAS číslo: 13201-46-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...