Tisk…

2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid 95+%

Dodavatel: Apollo Scientific

Synonyms: 3,5-Diiodotyrosine, H-DL-Tyr(3,5-DiI)-OH

OR27970-100MG OR27970-1G OR27970-250MG
APOSOR27970-100MGEA 2070 CZK
APOSOR27970-100MG APOSOR27970-1G APOSOR27970-250MG
2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid 95+%
2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Vzorec: C₉H₉I₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD00063076
CAS číslo: 66-02-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR